missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) sulfate pentahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 197720050

 View more versions of this product

Catalog No. AC197720050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper(II) sulfate pentahydrate
7758-99-8
Fine Crystals or Crystalline Powder
99% min. (Iodometry)
CuSO4·5H2O
3077
15, 2643
Solubility in water: 320g/L (20°C). Other solubilities: soluble in methanol,slightly soluble in alcohol and glycerol,insoluble in ethanol
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
249.68
CHEBI:31440
≥99%
Plastic bottle
110.0°C
Blue
5 kg
CuH10O9S
MFCD00149681
01,164; 02,89; 05,162; 06,141; 14,267
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper(2+) pentahydrate sulfate
24463
249.68
Analytical

Chemical Identifiers

7758-99-8
249.68
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
CuH10O9S
MFCD00149681
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
CHEBI:31440
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only