missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate solution, Volumetric, 0.1 M CuSO4, Honeywell Fluka™

Volumetric, 0.1 M CuSO4

$396.45 - $2068.15

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.602
MDL Number MFCD00010981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000451
View Documents
Honeywell Chemicals
351851L
1 L
Each for $396.45
Only null left
 
60046540
View Documents
Honeywell Chemicals
351856X1L
6 x 1 L
Case of 6 Each for $2,068.15
Only null left
 
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Chemical Identifiers

7758-98-7
159.602
ARUVKPQLZAKDPS-UHFFFAOYSA-L
24462
copper;sulfate
CuO4S
MFCD00010981
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
CHEBI:23414
[O-]S(=O)(=O)[O-].[Cu+2]

Specifications

7758-98-7
CuO4S
UN3082
ARUVKPQLZAKDPS-UHFFFAOYSA-L
copper;sulfate
24462
Poly bottle
1 L
MFCD00010981
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
[O-]S(=O)(=O)[O-].[Cu+2]
159.602
CHEBI:23414
Copper(II) sulfate solution
Videos
Safety and Handling

Safety and Handling

P273-P501

ShelfLife : 450 days from date of manufacture

DOTInformation : 9 / PGIII

Recommended Storage : Room Temp

SDS
missing translation for 'documents'

missing translation for 'documents'