missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper iodate, anhydrous, 95%, Thermo Scientific Chemicals
$121.71 - $2577.52
Chemical Identifiers
CAS | 13454-89-2 |
---|---|
Molecular Formula | CuI2O6 |
Molecular Weight (g/mol) | 413.35 |
MDL Number | MFCD00016055 |
InChI Key | LLVVIWYEOKVOFV-UHFFFAOYSA-L |
Synonym | copper iodate, copper 2+ diiodate, cupric iodate, copper iodate, anhydrous, cu.2io3 |
PubChem CID | 9801829 |
IUPAC Name | copper;diiodate |
SMILES | [Cu++].[O-][I](=O)=O.[O-][I](=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA8945904
|
Thermo Scientific Chemicals
08945904 |
2 g |
Each for $121.71
|
|
|||||
AA8945909
|
Thermo Scientific Chemicals
08945909 |
10 g |
Each for $579.14
|
|
|||||
AA8945918
|
Thermo Scientific Chemicals
08945918 |
50 g |
Each for $2,577.52
|
|
|||||
Description
It used as an intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13454-89-2 | |
413.35 | |
LLVVIWYEOKVOFV-UHFFFAOYSA-L | |
9801829 | |
[Cu++].[O-][I](=O)=O.[O-][I](=O)=O |
CuI2O6 | |
MFCD00016055 | |
copper iodate, copper 2+ diiodate, cupric iodate, copper iodate, anhydrous, cu.2io3 | |
copper;diiodate |
Specifications
(decomposition) | |
Green | |
2 g | |
CuI2O6 | |
copper iodate, copper 2+ diiodate, cupric iodate, copper iodate, anhydrous, cu.2io3 | |
LLVVIWYEOKVOFV-UHFFFAOYSA-L | |
copper;diiodate | |
9801829 | |
Hygroscopic | |
5.241^1^5 |
13454-89-2 | |
Powder | |
95% | |
MFCD00016055 | |
Soluble in water (1.36g/L). | |
[Cu++].[O-][I](=O)=O.[O-][I](=O)=O | |
413.35 | |
413.35 | |
Green powder | |
Copper iodate, anhydrous |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
DOTInformation : DOT Class: 5.1, Packing Group: II
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only