missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Oxide Wire, ACS Grade, 0.65 x 3mm, For Elementary Analysis, MilliporeSigma™

Supplier:  Technidata 1027650250

 View more versions of this product

Catalog No. M1027650250


Explore available promotions

Description

Description

For elementary analysis
Specifications

Specifications

Copper oxide
1317-38-0
250 g
MFCD00010979
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
[O--].[Cu++]
79.55
79.55. (molar mass)
Odorless
Dark gray wire (0.65 x 3mm)
Surface: CuO, Core: Cu2O
Wire
CuO
UN 3077
KKCXRELNMOYFLS-UHFFFAOYSA-N
copper(2+) oxidandiide
14829
ACS
Glass bottle

Chemical Identifiers

1317-38-0
79.55
KKCXRELNMOYFLS-UHFFFAOYSA-N
14829
[O--].[Cu++]
CuO
MFCD00010979
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
copper(2+) oxidandiide
Videos
Safety and Handling

Safety and Handling

H302: Harmful if swallowed.
H410: Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 215-269-1

SDS
Documents

Documents

Promotions

Promotions