missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis), Thermo Scientific Chemicals

$87.19 - $810.44

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1399018
View Documents
Thermo Scientific Chemicals
01399018
50 g
Each for $87.19
 
AA1399036
View Documents
Thermo Scientific Chemicals
01399036
500 g
Each for $103.77
 
AA13990A3
View Documents
Thermo Scientific Chemicals
013990A3
2 kg
Each for $239.96
 
AA13990A9
View Documents
Thermo Scientific Chemicals
013990A9
10 kg
Each for $810.44
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-50-8
63.55
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]

Specifications

7440-50-8
MFCD00010965
50 g
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
99%
Odorless
Powder
(metals basis)
Cu
UN3089
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
63.55
CHEBI:30052
−325 Mesh 10% max +325 Mesh
APS 8 to 11 Micron (FSSS)
Copper powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335-H500

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

EINECSNumber : 231-159-6

RTECSNumber : GL5325000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only