missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper shot, 99.5% (metals basis), Thermo Scientific Chemicals
$47.79 - $344.93
Chemical Identifiers
CAS | 7440-50-8 |
---|---|
Molecular Formula | Cu |
MDL Number | MFCD00010965 |
InChI Key | RYGMFSIKBFXOCR-UHFFFAOYSA-N |
Synonym | cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer |
PubChem CID | 23978 |
ChEBI | CHEBI:30052 |
IUPAC Name | copper |
SMILES | [Cu] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4421322
|
Thermo Scientific Chemicals
04421322 |
100 g |
Each for $47.79
|
|
|||||
AA4421336
|
Thermo Scientific Chemicals
04421336 |
500 g |
Each for $145.32
|
|
|||||
AA44213A3
|
Thermo Scientific Chemicals
44213A3 |
2 kg |
Each for $344.93
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-50-8 | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
CHEBI:30052 | |
[Cu] |
Cu | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
23978 | |
copper |
Specifications
7440-50-8 | |
99.5% (Metals basis) | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
[Cu] | |
63.55 | |
CHEBI:30052 | |
0 hPa | |
Copper |
Copper colored shot | |
Cu | |
100 g | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
copper | |
23978 | |
Odorless | |
Shot |
Safety and Handling
EINECSNumber : 231-159-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only