missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cork Borer Sets with Handles

Bore holes in corks and rubber stoppers.

$81.23 - $376.76

Specifications

Product Type Cork Borer
For Use With (Application) To bore holes in corks and Rubber stoppers
Includes Plated tubing, smooth Plastic handles
View More Specs

Products 4
Catalog Number Mfr. No. Diameter (English) Outer Diameter (Metric) Outer Quantity Price Quantity  
Catalog Number Mfr. No. Diameter (English) Outer Diameter (Metric) Outer Quantity Price Quantity  
S50166E
View Documents
Humboldt Inc
H9665
0.18 to 0.93 in. 4.8 to 23.8mm 15 Pack
Pack of 15 Each for $376.76
 
S50166C
View Documents
Customer Channels Group (Ccg)
H9663
0.18 to 0.65 in. 4.8 to 16.7mm 9 Pack
Pack of 9 Each for $248.00
 
S50166A
View Documents
Customer Channels Group (Ccg)
H9661
0.18 to 0.31 in. 4.8 to 7.8mm 3 Pack
Pack of 3 Each for $81.23
 
S50166D
View Documents
Customer Channels Group (Ccg)
H9664
0.18 to 0.79 in. 4.8 to 20.2mm 12 Pack
Pack of 12 Each for $317.15
 
Description

Description

  • Consist of plated tubing with precision-ground cutting edges
  • Smooth plastic handles
  • Handles are stamped with a number designation
  • Punch included with each set

Specifications

Specifications

Cork Borer
To bore holes in corks and Rubber stoppers
Plated tubing, smooth Plastic handles
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'