missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corn Oil, MP Biomedicals
Oleic Acid: 43-47%; Linoleic Acid: 39-42%; Palmitic Acid: 7-8%; Stearic Acid: 3-4%, Arachidic Acid: 0.5%; Approx. 7.6 lbs/gal
Supplier: MP Biomedicals Inc 0290141425
Specifications
Corn Oil | |
8001-30-7 | |
25 lb. |
Chemical Identifiers
8001-30-7 |
Safety and Handling
Recommended Storage : Room Temperature