Corning™ 0.05% Trypsin/0.53mM EDTA in HBSS w/o Calcium, Magnesium or Sodium Bicarbonate
Used to enzymatically release adherent cells from tissue culture plates for passaging.
Supplier:  Corning™ 25052CV
 View more versions of this product
Catalog No. MT25052CV
Explore available promotions
Description
Description
Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging.  Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action.  EDTA sequesters these ions and thus enhances the efficacy of trypsin.  This product contains trypsin at a concentration of 0.05% with 0.53 mM EDTA, and does not contain sodium bicarbonate.
Specifications
Specifications
-20°C
Cell Culture Dissociation Reagent
No Calcium, No Magnesium, No Sodium Bicarbonate
6 x 500 mL
Liquid
Sterile
7 to 7.6