missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ 384-Well Low-Volume Solid Microplates
Same low reagent volumes as 1536-well plates without the need for new instrumentation
Supplier: Corning™ 3824
Description
Polystyrene plates feature a unique square-to-round well design with a conical-shaped bottom that provides easier access and imaging for superior assay performance.
- Bottom reduces fluid and air entrapment, greater signal-to-noise ratio and reduced FP signal (mP) when working with very low volumes
- Each well has a raised rim to reduce cross-contamination during assay and liquid-handling process
- Choose black plates for fluorescence applications or white for luminescence applications
- Ideal for immunoassays
- Can bind medium (>10kDa) and large biomolecules that possess ionic groups and/or hydrophobic regions
- Working volume: 2 to 20μL
Specifications
No Barcode | |
Without Lid | |
White | |
384 | |
Non-sterile | |
Flat Bottom (F-well) | |
50/Cs. |
Standard | |
16 x 24 | |
Polystyrene | |
5 to 40 μL | |
Non-binding | |
Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process | |
White |