Corning™ 384-Well, Ultra-Low Binding, U-Shaped-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Sterile
Supplier:  Corning™ 4516
Catalog No. 12-456-722
Explore available promotions
Description
Description
With their novel and proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment (ULA) surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
  • Unique design shields each well to minimize well-to-well cross-talk.
  • You can culture and assay spheroids in the same microplate without the need for transfer to a new microplate.
Specifications
Specifications
Standard
0.06 cm2
5/Pk.
Spheroid Microplate
384
Sterile
90 μL
Closure
1/Pk.
With Lid
16 x 24
Black, Clear Bottom
Polystyrene
20 to 80 μL
Ultra-low Binding
Round Bottom (U-well)
5/Cs.
Product Suggestions
Product Suggestions