Learn More
Corning™ BioCoat™ Assay System
Automation-friendly design
$3831.63 - $3831.63
Specifications
| Color | Clear |
|---|---|
| Culture Area | 0.3 cm2 |
| Sterility | Non-sterile |
| Surface Treatment | Collagen |
| Well Count | 24 |
Includes: 24-Multiwell™ Insert plate, specialized media to perform 24 assays, feeder tray, and lid.
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
| Clear | |
| Non-sterile | |
| 24 | |
| 1 μm |
| 0.3 cm2 | |
| Collagen | |
| 24 multiwell insert plate, specialized media to perform 24 assays, feeder tray and lid |