missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ BioCoat™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate

Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:With Lid

Supplier:  Corning™ C4586

Catalog No. 15-100-164


Explore available promotions

Description

Description

High optical quality, glass bottom, black microplates are ideal for performing high content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.

  • High optical quality and scratch resistant glass
  • Glass bottom thickness of 200 μm is well suited for imaging microscopy.
  • Bottom flatness <50 μm to ensure planarity for imaging devices
  • Low background fluorescence and minimal cross-talk provides the highest possible optical quality for cell-based assays.
  • Half area 96-well microplate reduces reagent consumption.
Specifications

Specifications

Half-Area
8 x 12
96
Poly-D-Lysine
10/Cs.
With Lid
Black, Clear Bottom
Non-sterile
Flat Bottom (F-well)
1/Pk.
Videos
SDS
Documents

Documents

Promotions

Promotions