Corning™ Black 384-Well Polypropylene Assay Plate
Lower compound sticking and higher solvent resistance than polystyrene plates
Supplier:  Corning™ 3658
 View more versions of this product
Catalog No. 07-200-765
Explore available promotions
Description
Description
  • Ideal for fluorescence assays and for compound storage
  • Round-bottom wells
  • Untreated
Specifications
Specifications
Standard
16 x 24
Polypropylene
0.015 to 0.07 mL
Non-treated
For fluorescence assays and compound storage
Without Lid
Black
384
Non-sterile
Round
100/Cs.