missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Black and White 96-Well Polypropylene Assay Plates
Lower compound sticking and higher solvent resistance than polystyrene plates
$1672.51 - $1687.98
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Material | Polypropylene |
No. of Wells | 96 |
Volume (Metric) Working | 75 to 200 μL |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
07-200-761
|
Corning™
3355 |
White |
Case of 100 Each for $1,672.51
|
|
|||||
07-200-762
|
Corning™
3356 |
Black |
Case of 100 Each for $1,687.98
|
|
|||||
Description
- Ideal for fluorescence and luminescence assays and for compound storage
- Round-bottom wells
- Untreated surface
Specifications
Standard | |
Polypropylene | |
75 to 200 μL | |
Non-treated |
8 x 12 | |
96 | |
Non-sterile | |
Ideal for fluorescence and luminescence assays and for compound storage |