missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Cell Culture Treated Flasks

Manufactured from optically clear virgin polystyrene for optimum visibility and distortion-free microscopic viewing. Corning™ Cell Culture Treated Flasks are available in a variety of sizes, designs and cap styles to meet laboratory diagnostic needs.

$195.51 - $3514.53

Specifications

Closure Material HDPE
Material Polystyrene
For Use With (Application) For optimal cell attachment
Includes Lot numbers for ease in traceability
Shape Rectangular
View More Specs

Products 11
Catalog Number Mfr. No. Capacity (Metric) Closure Type Neck Style Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Type Neck Style Price Quantity  
10-126-64
View Documents
Corning™
431085
750 mL Phenolic Angled
Case of 50 Each for $1,322.57
 
10-126-60
View Documents
Corning™
431079
750 mL Plug Seal Angled
Case of 50 Each for $1,358.78
 
10-126-28 Edge
View Documents
Corning™
430639
70 mL Vented Canted
Case of 200 Each for $1,612.36
Edge
 
10-126-30 Edge
View Documents
Corning™
430168
70 mL Plug Seal Canted
Case of 500 Each for $3,514.53
Edge
 
10-126-32
View Documents
Corning™
430823
600 mL Plug Seal Canted
Case of 50 Each for $1,198.51
 
10-126-34
View Documents
Corning™
430825
600 mL Vented Canted
Case of 50 Each for $1,254.93
 
10-126-33
View Documents
Corning™
430824
600 mL Phenolic Canted
Case of 50 Each for $1,334.86
 
10-126-39
View Documents
Corning™
430372
70 mL Phenolic Canted
Case of 500 Each for $3,514.53
 
10-126-61
View Documents
Corning™
431080
750 mL Vented Angled
Case of 50 Each for $1,271.25
 
10-126-62
View Documents
Corning™
431081
1000 mL Plug Seal Angled
Case of 25 Each for $794.59
 
10-126-63
View Documents
Corning™
431082
1000 mL Vented Angled
Pack of 5 Each for $195.51
Case of 5 Pack for $977.55
 
Description

Description

  • Stackable, polystyrene
  • Cell culture surface treatment promotes cell attachment and growth
  • Tested for sterility, toxicity, cell attachment, and growth promotion
  • Optically clear surfaces provide distortion-free microscope viewing
  • Printed lot number for product traceability
  • Sterile and nonpyrogenic

Cap Types

  • Phenolic-style screw caps for cultures requiring gas exchange
  • Plug seal caps for airtight cultures
  • Vent caps to minimize cell culture contamination with gas exchange
Canted-neck flasks feature:
  • Antitip skirt
  • Molded graduations
  • Frosted marking areas on both sides
  • Support bar across the top
  • Redesigned neck on 75cm2 flask features redesigned neck to aid media pouring, 1.4 in. (3.6cm) height for lower stacking height; vent cap has Costar™-style concentric circle

Specifications

Specifications

HDPE
For optimal cell attachment
Rectangular
Yes
Polystyrene
Lot numbers for ease in traceability
Tissue Culture Treated
Sterile
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'