Corning™ Cell Lifter
Designed with a wide beveled edge for cell culture dishes. Corning™ Cell Lifters are an ideal tool for harvesting stem cells and colonies while maintaining high viability.
Supplier: Corning™ C3008
Description
Specifications
Sterile | |
7 in. | |
18 cm | |
Wide Beveled |
Cell culture dishes | |
0.74 in. | |
1.9 cm | |
Cell Lifter |