missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 10X
With low endotoxin content
Supplier: Corning™ 20030CV
Description
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.Specifications
2° to 30°C | |
Liquid | |
None, Magnesium | |
4.1 to 4.7 |
10 X | |
Dulbecco's Phosphate Buffered Saline | |
Calcium Chloride 1g/L, Potassium Chloride 2g/L, Potassium Dihydrogen Phosphate 2g/L, Magnesium Chloride Hexahydrate 1g/L, Sodium Chloride 80g/L, Disodium Phosphate 11.50g/L | |
6 x 500 mL |