Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X
With low endotoxin content.
Supplier:  Corning™ 21030CV
 View more versions of this product
Catalog No. MT21030CV
Explore available promotions
Description
Description
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time. This formulation contains calcium and magnesium.
Specifications
Specifications
Calcium and Magnesium
1 X
6 x 500 mL
6.9 to 7.1
2° to 30°C
DPBS
Calcium Chloride 0.10g/L, Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Magnesium Chloride Hexahydrate 0.1g/L, Sodium Chloride 8g/L, Disodium Phosphate Heptahydrate 2.1716g/L
Liquid