Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X
With low endotoxin content.
Supplier:  Corning™ 55031PB
 View more versions of this product
Catalog No. MT55031PB
Explore available promotions
Description
Description
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
Specifications
Specifications
No Calcium, No Magnesium
1 X
1 x 10,000 mL
Powder
2° to 8°C
DPBS
Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Sodium Chloride 8g/L, Disodium Phosphate 1.15g/L