Learn More
Corning™ Eagle's Minimum Essential Medium (MEM)
Used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts when supplemented with serum
Supplier: Corning™ 10009CV
Description
(This is suggested replacement for 10-009-CVR.)
[+] 1.5 g/L sodium bicarbonate, NEAA, L-glutamine, sodium pyruvate Technical Advantage: Product manufactured under industry-leading standards for low endotoxin content and superior results. Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. This research indicated that additions made to BME aided cell propagation. MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts. MEM 10-009-CV and -CM contain 1.5 g/L sodium bicarbonate, non-essential amino acids, L-glutamine, and sodium pyruvate.
- Complete Certificate of Analysis available for each production lot, along with full formulation information - Produced under the highest industry standards to ensure superior results Corning media certificates are available on our Resource Library.
Specifications
Red | |
Liquid | |
Sterile | |
7.2 |
2° to 8°C | |
Minimum Essential Medium Eagle's | |
Glutamine, Non Essential Amino Acids (NEAA), Sodium Bicarbonate | |
6 x 500 mL |