Learn More
Corning™ Lymphocyte Separation Medium
Designed for the in vitro isolation of lymphocytes from diluted whole blood.
$59.15 - $216.81
Specifications
| Form | Liquid |
|---|---|
| Product Type | Lymphocyte separation medium |
| Sterility | Sterile |
Description
Lymphocyte Separation Medium was originally designed for the in vitro isolation of lymphocytes from diluted whole blood. It is a sterile filtered, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/cm3 at 20°C.
Order Info
Packaged in PET bottles.
Specifications
| Liquid | |
| Lymphocyte separation medium | |
| Sterile |