missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Minimum Essential Medium Eagle (Mod.) 1X (MEM) with Glutamine

Guaranteed low endotoxin levels ≤0.025EU/mL

$160.41 - $291.44

Specifications

Content And Storage 2° to 8°C
Form Liquid
Product Type MEM Eagle/EBSS
Sterility Sterile
With Additives Glutamine, Earle's Balanced Salt Solution (EBSS)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MT10010CM
View Documents
Corning™
10010CM
6 x 1 L
Case of 6 Each for $291.44
 
MT10010CV
View Documents
Corning™
10010CV
6 x 500 mL
Case of 6 Each for $160.41
 
Description

Description

Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. This research indicated that additions made to BME aided cell propagation. MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts.  This formulation contains Earle's salts and L-glutamine.
Specifications

Specifications

2° to 8°C
MEM Eagle/EBSS
Glutamine, Earle's Balanced Salt Solution (EBSS)
Liquid
Sterile
7 to 7.4
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'