Corning™ IPTG, Powder
Produced under highest industry standards
Supplier:  Corning™ 46102RF
Catalog No. MT46102RF
Explore available promotions
Description
Description
IPTG (isopropylthiogalactoside)
  • Inducer of lac operon in bacteria
  • Used with X-gal for detecting lac gene activity
  • For screening of clones containing gene of interest in lac operon
Specifications
Specifications
For screening clones when used with X-Gal to detect lac gene activity
1000 mg