missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ CELLSTACK™ Culture Chambers

Featuring USP Class VI polystyrene construction for excellent optical clarity and mechanical strength. Corning™ CELLSTACK™ are available in five sizes and with two unique surface treatments in addition to the standard tissue culture treated surface.

Supplier:  Corning™ 3268

 View more versions of this product

Catalog No. 05-539-094


Explore available promotions

Description

Description

  • Two generous 26mm filling ports provide direct access to chamber bottom and flexibility for sterile filling and emptying
  • Standard 33mm threaded caps have 0.2μm pore nonwettable membranes sealed to caps—allowing gas exchange, preventing pressure buildup, and minimizing risk of contamination
  • Choice of surface treatments: standard Tissue Culture Treated Surface, CellBIND Surface and Ultra-Low Attachment Surface
  • Sterilized by gamma irradiation
  • Leak tested

  • Corning CellBIND™ surface treatment improves cell attachment leading to increased cell growth and yields and provides more consistent and even cell attachment. Surface helps adapt cells to reduced serum or serum-free conditions, requires no refrigeration or special handling and is stable at room temperature.
  • Ultra-Low attachment surface features a covalently bound hydrogel layer that effectively inhibits cellular attachment and minimizes protein absorption, enzyme activation and cellular activation

Specifications

Specifications

4.22 to 6.42 oz.
Orange
125 to 190 mL
Culture Chamber
636 cm²
One chamber
USP Class VI
33mm
Polystyrene
Yes
Tissue Culture-treated
Gamma Irradiated
Videos
SDS
Documents

Documents

Promotions

Promotions