Corning™ CellStripper Dissociation Reagent
Non-enzymatic cell dissociation solution gently dislodges adherent cells in culture
Supplier:  Corning™ 25056CI
Catalog No. MT25056CI
Explore available promotions
Description
Description
Designed to be an alternative to trypsin when used in conjunction with serum-free or serum-containing media
  • Formulated with a proprietary mixture of chelators
  • Reduces the risk of cell damage associated with protein digestive enzymes
Specifications
Specifications
2° to 8°C
Cell Culture Dissociation Reagent
6 x 100 mL
Liquid
Sterile