missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Cryogenic Vial Storage Boxes

For storage of internally and externally threaded vials in temperatures to –196°C (–320.8°F)

$457.13 - $610.85

Specifications

Color Orange
Material Polycarbonate
Shape Square
Type Cryo Storage Box
For Use With (Application) Storage of Internally and Externally Threaded Vials
View More Specs

Products 3
Catalog Number Mfr. No. Tube Array Height (English) Height (Metric) Holds Price Quantity  
Catalog Number Mfr. No. Tube Array Height (English) Height (Metric) Holds Price Quantity  
07-200-617
View Documents
Corning™
431121
10 x 10 2 in. 5.1 cm 100 (2 mL Internally Threaded Vial)
Case of 10 Each for $457.13
 
07-200-616
View Documents
Corning™
431120
9 x 9 3.75 in. 9.5 cm 81 (4 to 5 mL Internally or Externally Threaded Vial)
Case of 10 Each for $610.85
 
07-200-615
View Documents
Corning™
431119
9 x 9 2 in. 5.1 cm 81 (1 to 2 mL Internally or Externally Threaded Vial)
Case of 10 Each for $462.54
 
Description

Description

For storage of internally and externally threaded vials in temperatures to –196°C (–320.8°F)

  • Reusable racks are designed for use with most cryogenic vials
  • Cat. No. 431131 has a locking feature for use with all Corning self-standing vials
  • Cat. No. 431121 accepts internally threaded cryogenic vials only.
Specifications

Specifications

Orange
Square
Storage of Internally and Externally Threaded Vials
249.8°F
-320.8°F
5.25 in.
5.25 in.
Polycarbonate
Cryo Storage Box
Silk-Screened Lid
121°C
-196°C
13.3 cm
13.3 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'