Learn More
Corning™ DeckWorks™ Tip Stations, Nonsterile
Designed for single and multichannel pipetting stations
$175.67 - $186.27
Specifications
| Sterility | Non-sterile |
|---|---|
| Color | Natural |
| Tip Style | Round |
| Packaging | Multi-pack |
| For Use With (Equipment) | Universal Fit, Multichannel Pipetting Stations |
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|
10-320-731
|
Corning™
4142 |
1 to 200 μL |
Pack of 960 Each for $186.27
Case of 5 Pack for $931.35
|
|
|||||
|
10-320-730
|
Corning™
4141 |
0.1 to 10 μL |
Pack of 960 Each for $175.67
Case of 5 Pack for $878.35
|
|
|||||
Description
Corning DeckWorks tip station maximizes bench space while minimizing plastic waste. Tip Âstations contain 960 tips, are available in extended-length 10 μL and traditional 200 μL sizes, and are the ideal single and multi-channel pipetting stations. The 5-layer deck design (2 x 96) matches the microtiter plate format. This stable pipetting platform fits easily into sterile hood environments. Lids can be snap-positioned to expose a single side (96 tips) only. Applications include pre- and Âpost-PCR procedures, cell culture, and assays requiring large tip quantities. Corning DeckWorks tip stations are offered both sterile and nonsterile.
Specifications
| Non-sterile | |
| Round | |
| Universal Fit, Multichannel Pipetting Stations | |
| DeckWorks |
| Natural | |
| Multi-pack | |
| Yes | |
| Polypropylene |