missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Easy Grip Disposable Polystyrene Sterile Bottles
Use as replacement receivers for Corning Disposable Sterile Filter Systems or as storage bottles. Corning™ Disposable Sterile Bottles are easy to grip and ideal for storage of media, buffers and other aqueous solutions.
$256.78 - $595.20
Specifications
Disposable | Single-use |
---|---|
Material | Polystyrene |
Sterility | Sterile |
For Use With (Application) | Applications: Storage of media, buffers and other aqueous solutions |
Includes | Orange HDPE plug seal |
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
09-761-140
|
Corning™
431175 |
150 mL |
Case of 24 Each for $256.78
|
|
|||||
09-761-4
|
Corning™
430281 |
250 mL |
Case of 24 Each for $281.34
|
|
|||||
09-761-10
|
Corning™
430282 |
500 mL |
Case of 24 Each for $382.47
|
|
|||||
09-761-11
|
Corning™
430518 |
1 L |
Case of 24 Each for $595.20
|
|
|||||
Description
- Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
- Corning low profile style has easy grip sides that facilitate handling
- Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
- Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
- Sterile
- Nonpyrogenic
Specifications
Single-use | |
Sterile | |
Orange HDPE plug seal | |
24/Cs. | |
24 Case |
Polystyrene | |
Applications: Storage of media, buffers and other aqueous solutions | |
4.5 cm | |
2/Pk. | |
Graduated Easy Grip Storage Bottle |