Corning™ DMSO-Resistant Lid for Low Fluorescence Assay Plates
Robotics-friendly, nonsterile
Supplier:  Corning™ 3085
Catalog No. 07-201-200
Explore available promotions
Description
Description
  • All lids are made of rigid polystyrene except where indicated.
  • All lids have a corner notch on the A1 corner (except where indicated) to correspond to the corner notches found on all Corning™ microplates.
  • The universal lid without a corner notch (Cat. No. 3098) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids.
  • The black universal lid (Cat. No. 3935) is suitable for fluorescent and other light-sensitive assays.
  • The DMSO-resistant cyclic olefin copolymer (COC) lid (Cat. No. 3085) is tinted amber in color for light-sensitive assays and is 100% DMSO-resistant.
  • Specifications
    Specifications
    Cycloolefin
    50/Cs.
    Resistant Lid
    Non-sterile
    25/Pk.
    Low Fluorescence Assay Plate