missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Fibronectin, Human

Promotes cellular attachment, spreading, and proliferation on tissue-culture surfaces. Corning™ Fibronectin, Human can be used as an additive to serum-free culture media.

$314.38 - $1074.33

Specifications

Content And Storage 2° to 8°C
Product Type Fibronectin
pH 11
Source Human Plasma
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CB-40008A
View Documents
Corning™
356008
5 mg
Each for $1,074.33
Only null left
 
CB-40008
View Documents
Corning™
354008
1 mg
Each for $314.38
Only null left
 
Description

Description

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

The principal functions of fibronectin appear to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis.

Specifications

Specifications

2° to 8°C
Fibronectin
11
Human Plasma
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'