Corning™ LSE™ Vortex Mixer
Variable speed mixer provides fast, efficient mixing with minimal vibration
Supplier:  Corning™ C6775
Catalog No. 10-320-807
Explore available promotions
Alert:
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Includes: Cup head for all standard microtubes and centrifuge tubes
Description
Description
The Corning LSE variable speed vortex mixer provides fast, efficient mixing with minimal vibration. The unit features a powerful, counter-balanced motor drive, and continuous or 'touch' mix modes. A range of accessories are available for all common sample tubes and containers.
  • Variable speed for gentle to vigorous mixing
  • Optimized counter balance for minimal vibration
  • Lightweight and portable
  • Includes cup head for all standard microtubes and centrifuge tubes
  • Mix a variety of tubes and plates with optional accessory heads
  • Touch and continuous mix modes
Specifications
Specifications
0 to 3400 rpm
120 V
50/60 Hz
13 cm
14 cm
16 cm
2.2 kg
120 V, 50/60 Hz
Vortex Mixer
5.11 in.
5.51 in.
6.29 in.
4.8 lb.
Videos
SDS
Documents
Documents
Accessories