Learn More
Corning™ Matrigel™ hESC-Qualified Matrix
Compatible with mTeSR™1-devloped by StemCell Technologies. Corning™ Matrigel™ hESC-Qualified Matrix is widely accepted as an alternative substrate for the culture of hES cells as well as human induced pluripotent stem (iPS) cells.
Supplier: Corning™ 354277
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Recommended Storage
Store at -20°C. Store aliquots in either the -20°C or -70°C freezer until ready for use. Freeze thaws should be minimized by aliquotting into one time use aliquots. Do not store in frost-free freezer. Keep frozen.
Specifications
Store at -20°C. Store aliquots in either the -20°C or -70°C freezer until ready for use. Freeze thaws should be minimized by aliquotting into one time use aliquots. Do not store in frost-free freezer. Keep frozen. | |
hESC-qualified Matrix | |
5 mL |
For research use only.