Learn More
Corning™ Matrigel™ Matrix
Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.
$505.06 - $953.69
Specifications
Content And Storage | -20°C |
---|---|
Product Type | Basement Membrane Matrix |
For Use With (Application) | Cell growth and differentiation; metabolism/toxicology studies; invasion assays; in vitro and in vivo angiogenesis assays; in vivo angiogenesis studies and augmentation of tumors in immunosuppressed mice |
Product Line | Matrigel |
Source | Engelbreth-Holm-Swarm Mouse Tumor |
Alert:
Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer
Catalog Number | Mfr. No. | Quantity | Without Additives | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Without Additives | Price | Quantity | |||||
CB-40234A
|
Corning™
356234 |
5 mL | No LDEV |
Each for $505.06
|
|
|||||
CB-40234C
|
Corning™
356237 |
10 mL | No LDEV, No Phenol Red |
Each for $953.69
|
|
|||||
CB-40234
|
Corning™
354234 |
10 mL | No LDEV |
Each for $815.31
|
|
|||||
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
-20°C | |
Cell growth and differentiation; metabolism/toxicology studies; invasion assays; in vitro and in vivo angiogenesis assays; in vivo angiogenesis studies and augmentation of tumors in immunosuppressed mice | |
Engelbreth-Holm-Swarm Mouse Tumor |
Basement Membrane Matrix | |
Matrigel |