Learn More
Corning™ Matrigel™ GFR Basement Membrane Matrix
Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ GFR Basement Membrane Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.
$638.15 - $1,012.73
Specifications
| Product Type | GFR Basement Membrane Matrix |
|---|---|
| For Use With (Application) | Suitable for applications that require a highly defined basement membrane preparation. |
| Source | Engelbreth-Holm-Swarm Mouse Tumor |
| Content And Storage | -20°C |
Alert:
Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer
Description
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Specifications
| GFR Basement Membrane Matrix | |
| Suitable for applications that require a highly defined basement membrane preparation. | |
| Engelbreth-Holm-Swarm Mouse Tumor | |
| -20°C |