Corning™ Multiple Position Stirrers
Stir up to nine vessels at once
Supplier:  Corning™ 440825
 View more versions of this product
Catalog No. 11-497-2
Explore available promotions
Description
Description
This Corning™ five position stirrer has integral controls that can accommodate up to five 400 mL vessels. It operates on 120V/60 Hz.
  • Compact design saves valuable bench space
  • Offers powerful stirring from 350 to 2000 rpm.
  • Rugged lightweight glass filled resin case
  • Two-year warranty
Specifications
Specifications
350 to 2000 rpm
400 mL
130 mm
60 Hz
5
120 V
120V 60Hz
MP5I
Videos
SDS
Documents
Documents
Accessories