Corning™ Polypropylene Plates
Manufactured from polypropylene for optimum visibility and high chemical resistance. Corning™ Polypropylene Plates are an excellent choice for transport and long-term storage.
Supplier:  Corning™ 3359
 View more versions of this product
Catalog No. 07-200-745
Explore available promotions
Description
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile
Specifications
Specifications
Standard
8 x 12
Polypropylene
Sterile
Round
100/Cs.
100/Cs.
Without Lid
Clear
96
Non-treated
Transport and longterm storage
25/Pk.