missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Primaria™ Cell Culture Multiwell™ Plates

Features a surface treatment that is a permanent, stable modification of the polystyrene surface incorporating a mixture of anionic and cationic functional groups. Corning™ Primaria™ Cell Culture Multiwell™ Plates feature a flat bottom with low evaporation lid.

Supplier:  Corning™ 353847

Catalog No. 08-772-4G


Only null left
Explore available promotions

Description

Description

  • Unique nitrogen-containing tissue culture surface chemistry
  • Improves attachment, spreading, and growth for many primary cells or cell lines
  • Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
  • Stable, permanent surface modification
  • Optically clear
  • No special storage required
  • Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0.1 EU/mL
Specifications

Specifications

4 x 6
Yes
Flat
Clear
Polystyrene
Sterile
Cell Culture Plate
24
Lid
1/Pk.
50/Cs.
85.47 mm
Clear
Polystyrene
1 mL (Well)
2 cm2
Rectangular
Primaria
1.5 to 2.2 mL
Suitable for growing a variety of primary cells and cell lines that grow poorly on conventional, negatively charged, standard tissue culture surfaces
50/Cs.
Individually Wrapped
127.63 mm
21.43 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions