Learn More
Corning™ Primaria™ Tissue Culture Flasks
Improves attachment, spreading and growth for many primary cells or cell lines. Corning™ Primaria™ Tissue Culture Flasks are suitable for growing cells that grow poorly on conventional, negatively charged, standard tissue culture surfaces—including tumor cells, endothelial cells and hepatocytes.
$794.15 - $1559.56
Specifications
Closure Color Blue
Closure Material Phenolic
Material Polystyrene
Shape Rectangular
Sterility Sterile
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Culture Area Closure Type Neck Style Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Culture Area Closure Type Neck Style Price Quantity  
08-772
View Documents
Corning™
353824
250 mL 75 cm2 Plug Seal Straight
Case of 100 Each for $1,221.04
Only null left
08-772-45
View Documents
Corning™
353808
50 mL 25 cm2 Vented Canted
Case of 100 Each for $794.15
Only null left
08-772-46
View Documents
Corning™
353810
250 mL 75 cm2 Vented Straight
Case of 100 Each for $1,516.52
Only null left
08-772-4E
View Documents
Corning™
353813
50 mL 25 cm2 Plug Seal Canted
Case of 200 Each for $1,559.56
Only null left
Description
Description
  • Unique nitrogen-containing tissue culture surface chemistry
  • Improves attachment, spreading, and growth for many primary cells or cell lines
  • Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
  • Stable, permanent surface modification
  • Optically clear
  • No special storage required
  • Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0.1 EU/mL
Order Info
Smaller flasks packed 20 per sleeve in cases of 200 (08-772-45 in cases of 100); larger flasks packed five per sleeve in cases of 100
Specifications
Specifications
Blue
Polystyrene
Sterile
Phenolic
Rectangular
Rectangular
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.