Corning™ PYREX™ VISTA™ Class A Volumetric Flask
Necks are tooled for standard taper glass stoppers.
$245.31 - $245.31
Specifications
Closure Material | Glass |
---|---|
Material | Glass, Borosilicate Glass |
Neck Style | Long |
Graduated | Yes |
Bottom Shape | Flat |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
S76109J
|
Corning™
706402LEM |
2000 mL |
Each for $245.31
Case of 2 Each for $490.62
|
|
|||||
Description
- Provide precise volume measurement
- Graduation line is sharp and permanent
- Large blue block letters make the inscription easy to read
Compliance
Manufactured to Class A tolerances as established by ASTM™ E-694 for volumetric ware, ASTM E-542 for calibration of volumetric ware, and ASTM E-288 for volumetric flasks.
Specifications
Glass | |
Long | |
Flat |
Glass, Borosilicate Glass | |
Yes |