Corning™ PYREX™ Watch Glasses
Heavy walls and uniform fire-polished edges for strength.
Supplier:  Corning™ 998590EMD
 View more versions of this product
Catalog No. S34820
Explore available promotions
Description
Description
Feature a high chemical durability which prevents contamination.
Specifications
Specifications
Reusable
Round
Used as Beaker Covers
Plain
9 cm
Borosilicate Glass
Watch Glass
For 400mL beaker
3.54 in.