Corning™ Reusable Phenolic Screw Caps With Rubber Liners
Specially formulated phenolic resin resists the effects of temperature and steam in autoclaving
Supplier:  Corning™ 999928
 View more versions of this product
Catalog No. 02-883-9B
Explore available promotions
Description
Description
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
Specifications
Specifications
White
144/Cs.
Black
Rubber Liner
Phenolic Resin
144/Pk.
Phenolic