Corning™ Reusable Phenolic Screw Caps With Rubber Liners
Specially formulated phenolic resin resists the effects of temperature and steam in autoclaving
Supplier:  Corning™ 9999241
 View more versions of this product
Catalog No. 02-883-9E
Explore available promotions
Description
Description
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
Specifications
Specifications
Black
Phenolic Resin
36/Pk.
24-410
Phenolic
18.5 mm
144/Cs.
Black
Rubber
Rubber Liner