Corning™ Reusable Plastic Erlenmeyer Flasks with Blue Graduations
Translucent polypropylene; with polypropylene screw cap.
Supplier:  Corning™ 4985P500
 View more versions of this product
Catalog No. 07-202-113
Explore available promotions
Description
Description
Translucent polypropylene flasks are manufactured with blue graduations for reference. The flasks have a screw neck and are supplied with a PP cap.
Specifications
Specifications
Polypropylene
Polypropylene
Narrow Mouth
1/Pk.
500 mL
100 mL
6/Cs.
Screw Cap