Corning™ Reusable Plastic Erlenmeyer Flasks with Red Graduations
Translucent polymethylpentene; with polypropylene screw cap
Supplier:  Corning™ 4990P100
 View more versions of this product
Catalog No. 07-202-116
Explore available promotions
Description
Description
Translucent polymethylpentene flasks are manufactured with red graduations for reference. The flasks have a screw neck and are supplied with a PP cap.
Specifications
Specifications
Polypropylene
Polypropylene
Narrow Mouth
1/Pk.
100 mL
20 mL
6/Cs.
Screw Cap
Product Suggestions
Product Suggestions