Corning™ PYREX™ Plain Bore Rotaflo™ Stopcock
Combined PTFE and borosilicate glass forms a leak- and freeze-resistant seal
Supplier: Corning™ 747010
Description
These high performance Rotaflo™ stopcocks can be adjusted. They are designed so that only chemically resistant polytetrafluoroethylene (PTFE) and PYREX™ borosilicate glass are in contact with liquids and gases. The Rotaflo™ stopcock design permits fine flow adjustment. Lubrication is not required and assembly is fast because there are no O-Rings, washers or nuts. The seal is visible when open or closed.
Specifications
High Vacuum Stopcock | |
High Vacuum | |
0.5 in. | |
8.8 in. | |
-20°C to 200°C | |
1/Pk. |
0 to 10 mm | |
Glass | |
13 mm | |
224 mm | |
-4°F to 392°F | |
1/Cs. |