Corning™ PYREX™ Plain Bore Rotaflo™ Stopcock
Combined PTFE and borosilicate glass forms a leak- and freeze-resistant seal
Supplier: Corning™ 74733
Description
These high performance Rotaflo™ stopcocks can be adjusted. They are designed so that only chemically resistant polytetrafluoroethylene (PTFE) and PYREX™ borosilicate glass are in contact with liquids and gases. The Rotaflo™ stopcock design permits fine flow adjustment. Lubrication is not required and assembly is fast because there are no O-Rings, washers or nuts. The seal is visible when open or closed.
Specifications
High Vacuum Stopcock | |
Plain Bore, Two-Way, 90° Angle | |
0.31 in. | |
6.1 in. | |
-20°C to 200°C | |
1/Pk. |
0 to 3 mm | |
Glass | |
8 mm | |
155 mm | |
-4°F to 392°F | |
1/Cs. |