Corning™ PYREX™ Plain Bore Rotaflo™ Stopcock
Combined PTFE and borosilicate glass forms a leak- and freeze-resistant seal
Supplier:  Corning™ 74753
 View more versions of this product
Catalog No. 14-630-30A
Explore available promotions
Description
Description
These high performance Rotaflo™ stopcocks can be adjusted. They are designed so that only chemically resistant polytetrafluoroethylene (PTFE) and PYREX™ borosilicate glass are in contact with liquids and gases. The Rotaflo™ stopcock design permits fine flow adjustment. Lubrication is not required and assembly is fast because there are no O-Rings, washers or nuts. The seal is visible when open or closed.
Specifications
Specifications
High Vacuum Stopcock
Plain Bore, Three-Way
0.31 in.
8.4 in.
-20°C to 200°C
1/Pk.
0 to 3 mm
Glass
8 mm
215 mm
-4°F to 392°F
1/Cs.