Corning™ Thermowell™ GOLD Polypropylene PCR Tubes
Designed for precise fit in heat blocks
Supplier:  Corning™ 3750
 View more versions of this product
Catalog No. 07-200-396
Explore available promotions
Description
Description
Individual PCR tubes are made of thin wall polypropylene and designed for precise fit in heat blocks to optimize heat transfer. Tubes are RNase-/DNase-free, are autoclavable at 121°C, and withstand centrifugation to 10,000 x g.
Specifications
Specifications
RNase, DNase-free
Clear
500/Pk.
Heat blocks to optimize heat transfer
Non-sterile
10,000 x g
Autoclavable
Thin Wall PCR Tube
Clear
0.5 mL
Polypropylene
Flat
Polypropylene
Flat
Tube