missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Clear Bottom Black or White Polystyrene Microplates permit direct microscopic viewing. For fluorescent and luminescent assays; ideal for performing cell-based and molecular assays.
$2689.34 - $3186.69
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
Color | Black, Clear Bottom |
Material | Polystyrene |
Catalog Number | Mfr. No. | Color | Surface Treatment | No. per Case | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Surface Treatment | No. per Case | Price | Quantity | |||
07-200-567
|
Corning™
3631 |
Black, Clear Bottom | Non-treated | 100 |
Case of 100 Each for $2,689.34
|
|
|||
07-200-625
|
Corning™
C3601 |
Black, Clear Bottom | High Binding | 100 |
Case of 100 Each for $3,186.69
|
|
|||
Description
- Molded polystyrene with opaque walls and optically clear bottoms
- Bottoms are 60% thinner than conventional polystyrene microplates for lower background flourecence and enabling readings down to 340nm
- Opaque black or white walls prevent well to well crosstalk
- Recommended working volumes of 75 to 200μL
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
- Plate dimensions meet proposed industry standards
Order Info
Cat. No. 07-200-568 also available in a pack of 25.
Specifications
Standard | |
8 x 12 | |
Polystyrene | |
75 to 200 μL | |
Flat Bottom (F-well) | |
100/Cs. |
Without Lid | |
Black, Clear Bottom | |
96 | |
Non-sterile | |
100 |